Learn More
Contec™ VertiKlean™ MAX™ Hands Free Mop Removal Tool
BIOPHARMAGREENER_CHOICE
Catalog No. 19168542 Shop All Contec Inc. Products
Change view
Click to view available options
Material:
Stainless Steel (304)
Catalog No. Material
19-168-542 Stainless Steel (304)
1 options
This item is not returnable. View return policy
Catalog No. 19-168-542 Supplier Contec™ Supplier No. 2639
Add to Cart
Add to Cart
This item is not returnable. View return policy
Hands-free stainless-steel mop removal tool for Compact Bucket System
Allows for hands free mop removal and disposal of all verisons of Contec's VertiKlean™ MAX™ and VertiKlean™ MAX™ Sealed Edge style mops
Specifications
Product Type Mop Head Removal Tool, Waste Bag Holder
Sterility Non-sterile
Quantity 1/Cs.
For Use With (Equipment) VertiKlean™ MAX™ and VertiKlean™ MAX™ Sealed Edge Mops
Material Stainless Steel (304)