Learn More
MILLER STEPHENSON CHEMICAL COMPANY INC VERTREL MCA CLEANING AGENT 1GA
SDP
Catalog No. NC9688901
Encompass_Preferred
Vertrel MCA Cleaning Agent ,1Gal,A nonflammable,azeotrope of Vertrel XF with trans-1,2-dichloroethylene. Its enhanced solvency power,compared to Vertrel XF,makes it particularly effective for difficult soil conditions
Catalog No. NC9688901 Supplier MILLER STEPHENSON CHEMICAL COMPANY INC Supplier No. MS7521GAL
Vertrel MCA Cleaning Agent ,1Gal,A nonflammable,azeotrope of Vertrel XF with trans-1,2-dichloroethylene. Its enhanced solvency power,compared to Vertrel XF,makes it particularly effective for difficult soil conditions