Learn More
MILLER STEPHENSON CHEMICAL COMPANY INC VERTREL XF CLEANING AGENT 1GAL
SDP
Catalog No. NC9688899
Encompass_Preferred
Vertrel XF Cleaning agent,1Gal,A very mild,nonflammable solvent. Excellent compatibility with most plastics,Applications :cleaning and rinsing,drying,particulate removal,fluorocarbon lubricant carrier,heat transfer and replacement for many
Catalog No. NC9688899 Supplier MILLER STEPHENSON CHEMICAL COMPANY INC Supplier No. MS7821GAL
Vertrel XF Cleaning agent,1Gal,A very mild,nonflammable solvent. Excellent compatibility with most plastics,Applications :cleaning and rinsing,drying,particulate removal,fluorocarbon lubricant carrier,heat transfer and replacement for many