Learn More
MILLER STEPHENSON CHEMICAL COMPANY INC VERTREL XF CLEANING AGENT 1QT
SDP
Catalog No. NC9715008
Encompass_Preferred
Vertrel XF Cleaning agent,1 QT,A very mild,nonflammable solvent,Excellent compatibility with most plastics,Applications :cleaning and rinsing,drying,particulate removal,fluorocarbon lubricant carrier,heat transfer and replacement for many
Catalog No. NC9715008 Supplier MILLER STEPHENSON CHEMICAL COMPANY INC Supplier No. MS782QT
Vertrel XF Cleaning agent,1 QT,A very mild,nonflammable solvent,Excellent compatibility with most plastics,Applications :cleaning and rinsing,drying,particulate removal,fluorocarbon lubricant carrier,heat transfer and replacement for many