Learn More
Medchemexpress LLC HY-12098 1mg Medchemexpress, Verubulin (hydrochloride) CAS:917369-31-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY120981MG
Encompass_Preferred
Medchemexpress, HY-12098 1mg Verubulin (hydrochloride) CAS:917369-31-4 Verubulin hydrochloride (MPC-6827 hydrochloride) is a blood brain barrier permeable microtubule -disrupting agent, with potent and broad-spectrum in vitro and in vivo cytotoxic activities. Verubulin hydrochloride (MPC-6827 hydrochloride) exhibits potent anticancer activity in human MX-1 breast and other mouse xenograft cancer models. Verubulin hydrochloride (MPC 6827 hydrochloride) is a promising candidate for the treatment of multiple cancer types [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2555