Learn More
Selleck Chemical LLC Verubulin hydrochloride S3374-5mg
SDP
Catalog No. 503108328
Encompass_Preferred
Verubulin hydrochloride(MPC 6827 hydrochloride) is a microtubule destabilizer and vascular disrupting agent that achieve high brain concentration relative to plasma in animals
Catalog No. 50-310-8328 Supplier Selleck Chemical LLC Supplier No. S33745MG
Verubulin hydrochloride(MPC 6827 hydrochloride) is a microtubule destabilizer and vascular disrupting agent that achieve high brain concentration relative to plasma in animals