Learn More
Medchemexpress LLC Verubulin hydrochloride | 917369-31-4 | 98.7% | 1 ML
SDP
Catalog No. 5000232728
Encompass_Preferred
Verubulin hydrochloride is the hydrochloride salt of verubulin (MPC-6827), a microtubule/tubulin inhibitor used in preclinical research. It is supplied as a ready-to-use 10 mM solution in DMSO and is also available as solid quantities for custom preparation. Intended for research use only.
  • Highly pure product (98.7% typical).
  • Provided as 10 mM, 1 mL solution in DMSO for ready-to-use applications.
  • Available as solid in multiple masses (mg scale) for flexible dosing.
  • Molecular weight 315.8 g/mol; formula C17H18ClN3O.
  • Suitable for in vitro and in vivo preclinical research; not for human use.
Catalog No. 50-002-32728 Supplier Medchemexpress LLC Supplier No. HY1209810MM 1ML
Verubulin hydrochloride is the hydrochloride salt of verubulin (MPC-6827), a microtubule/tubulin inhibitor used in preclinical research. It is supplied as a ready-to-use 10 mM solution in DMSO and is also available as solid quantities for custom preparation. Intended for research use only.
  • Highly pure product (98.7% typical).
  • Provided as 10 mM, 1 mL solution in DMSO for ready-to-use applications.
  • Available as solid in multiple masses (mg scale) for flexible dosing.
  • Molecular weight 315.8 g/mol; formula C17H18ClN3O.
  • Suitable for in vitro and in vivo preclinical research; not for human use.