Learn More
Medchemexpress LLC Verubulin hydrochloride | 917369-31-4 | 98.7% | 10 MG
SDP
Catalog No. 5000231959
Encompass_Preferred
Verubulin hydrochloride is the hydrochloride salt of verubulin, a blood-brain barrier-permeable microtubule-disrupting agent used in research for studies of tubulin polymerization, microtubule dynamics, and anticancer pharmacology. It is supplied as a high-purity research reagent for in vitro and in vivo studies.
  • Blood-brain barrier permeable microtubule disruptor.
  • Potent, broad-spectrum cytotoxic and antitumor activity.
  • Hydrochloride salt form.
  • Available in multiple pack sizes, including 10 mg.
  • High purity suitable for research applications.
Catalog No. 50-002-31959 Supplier Medchemexpress LLC Supplier No. HY1209810MG
Verubulin hydrochloride is the hydrochloride salt of verubulin, a blood-brain barrier-permeable microtubule-disrupting agent used in research for studies of tubulin polymerization, microtubule dynamics, and anticancer pharmacology. It is supplied as a high-purity research reagent for in vitro and in vivo studies.
  • Blood-brain barrier permeable microtubule disruptor.
  • Potent, broad-spectrum cytotoxic and antitumor activity.
  • Hydrochloride salt form.
  • Available in multiple pack sizes, including 10 mg.
  • High purity suitable for research applications.