Learn More
Medchemexpress LLC Verubulin hydrochloride | 917369-31-4 | 98.7% | 100 MG
SDP
Catalog No. 5000232798
Encompass_Preferred
Verubulin hydrochloride is the hydrochloride salt of verubulin (MPC-6827), a blood-brain barrier permeable microtubule-disrupting agent with demonstrated in vitro and in vivo anticancer activity. It is supplied for research use in solid and solution formats.
  • Blood-brain barrier permeable microtubule disruptor.
  • Demonstrated cytotoxic activity in MX-1 breast and other xenograft models.
  • Available as solid and as solution in DMSO; pack sizes typically range from 5 mg to 500 mg.
  • High purity (approx. 98.7%).
  • Molecular formula C17H18ClN3O; molecular weight 315.8 g/mol.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-32798 Supplier Medchemexpress LLC Supplier No. HY12098100MG
Verubulin hydrochloride is the hydrochloride salt of verubulin (MPC-6827), a blood-brain barrier permeable microtubule-disrupting agent with demonstrated in vitro and in vivo anticancer activity. It is supplied for research use in solid and solution formats.
  • Blood-brain barrier permeable microtubule disruptor.
  • Demonstrated cytotoxic activity in MX-1 breast and other xenograft models.
  • Available as solid and as solution in DMSO; pack sizes typically range from 5 mg to 500 mg.
  • High purity (approx. 98.7%).
  • Molecular formula C17H18ClN3O; molecular weight 315.8 g/mol.
  • For research use only; not for human or veterinary use.