Learn More
Medchemexpress LLC Verubulin hydrochloride | 917369-31-4 | 98.7% | 50 MG
SDP
Catalog No. 5000231960
Encompass_Preferred
Verubulin hydrochloride is the hydrochloride salt of verubulin (MPC-6827), a quinazoline-derived microtubule polymerization inhibitor with reported antitumor activity. It is supplied as a solid research reagent for preclinical studies investigating microtubule dynamics and anticancer mechanisms.
  • Hydrochloride salt suitable for enhanced solubility in polar solvents.
  • Potent microtubule polymerization inhibitor used in anticancer research.
  • Reported to be brain-penetrant, enabling neurobiology applications.
  • High purity typical for analytical and preclinical work (98.7%).
  • Stable as a solid at 4°C; follow recommended storage conditions for solutions.
Catalog No. 50-002-31960 Supplier Medchemexpress LLC Supplier No. HY1209850MG
Verubulin hydrochloride is the hydrochloride salt of verubulin (MPC-6827), a quinazoline-derived microtubule polymerization inhibitor with reported antitumor activity. It is supplied as a solid research reagent for preclinical studies investigating microtubule dynamics and anticancer mechanisms.
  • Hydrochloride salt suitable for enhanced solubility in polar solvents.
  • Potent microtubule polymerization inhibitor used in anticancer research.
  • Reported to be brain-penetrant, enabling neurobiology applications.
  • High purity typical for analytical and preclinical work (98.7%).
  • Stable as a solid at 4°C; follow recommended storage conditions for solutions.