Learn More
Medchemexpress LLC HY-14875 5mg Medchemexpress, Verucerfont CAS:885220-61-1 Purity:>98%
SDP
Catalog No. 502081652
Encompass_Preferred
Medchemexpress, HY-14875 5mg Verucerfont CAS:885220-61-1 Verucerfont is a corticotropin-releasing factor receptor 1 (CRF1) antagonist with IC50s of ~6.1, >1000 and >1000 nM for CRF1, CRF2, and CRF-BP, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1652 Supplier Medchemexpress LLC Supplier No. HY148755MG
Medchemexpress, HY-14875 5mg Verucerfont CAS:885220-61-1 Verucerfont is a corticotropin-releasing factor receptor 1 (CRF1) antagonist with IC50s of ~6.1, >1000 and >1000 nM for CRF1, CRF2, and CRF-BP, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.