Learn More
Medchemexpress LLC HY-P2657 50ug Medchemexpress, Verucopeptin CAS:138067-14-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP265750UG
Encompass_Preferred
Medchemexpress, HY-P2657 50ug Verucopeptin CAS:138067-14-8 Verucopeptin is a potent HIF-1 (IC50=0.22 μM) inhibitor and decreases the expression of HIF-1 target genes and HIF-1α protein levels. Verucopeptin strongly inhibits v-ATPase activity by directly targeting the v-ATPase ATP6V1G subunit but not ATP1V1B2 or ATP6V1D. Verucopeptin exhibits antitumor activity against multidrug resistance (MDR) cancers and can be used for cancer research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3090