Learn More
Electron Microscopy Sciences SEM Very High Resolution Gold on Carbon Test Specimen, JEOL Stub
SDP
Catalog No. 5029612
Encompass_Preferred
For very high resolution performance testing this specimen has a smaller gold particle size ranging from 2 nm to 30 nm as compared to the high resolution test above. Ideal for testing at instrument magnifications of 50,000x and above.
Mount: JEOL Stub
Catalog No. 50-296-12 Supplier Electron Microscopy Sciences Supplier No. 7951102
For very high resolution performance testing this specimen has a smaller gold particle size ranging from 2 nm to 30 nm as compared to the high resolution test above. Ideal for testing at instrument magnifications of 50,000x and above.
Mount: JEOL Stub