Learn More
Bachem Fmoc-Glu(Cys(Fmoc)-Gly-OH)-OH
SDP
Supplier:  Bachem 4026654.0250
Encompass_Preferred
N,S-Bis-Fmoc-glutathione, Fmoc-Glu(Cys(Fmoc)-Gly-OH)-OH, 149438-56-2, C40H37N3O10S, Mr 751.81, 250mg. Very potent inhibitor of mammalian glyoxalase II (Ki = 0.08 uM for glyoxalase II purified from calf liver).
Catalog No. 50-194-7050