Learn More
Medchemexpress LLC (±)-Vesamicol hydrochloride | 120447-62-3 | 99.8% | 25 MG
SDP
Catalog No. 5000414388
Encompass_Preferred
(±)-Vesamicol hydrochloride ((±)-AH5183 hydrochloride) is a potent vesicular acetylcholine transport inhibitor with a Ki of 2 nM. It also displays high affinity for σ1 and σ2 receptors with Kis of 26 nM and 34 nM, respectively. The pharmacological actions of (±)-Vesamicol are attributed to the inhibition of acetylcholine transport into synaptic vesicles and subsequent quantal release of acetylcholine.
  • Potent vesicular acetylcholine transport inhibitor
  • Displays high affinity for σ1 and σ2 receptors
  • Inhibits acetylcholine transport into synaptic vesicles
  • Reduces subsequent quantal release of acetylcholine
  • Increases cytosolic acetylcholine levels in rat brain
  • Decreases vesicular acetylcholine levels in rat brain (except striatum)
Catalog No. 50-004-14388 Supplier Medchemexpress LLC Supplier No. 25MGHYB1813A
(±)-Vesamicol hydrochloride ((±)-AH5183 hydrochloride) is a potent vesicular acetylcholine transport inhibitor with a Ki of 2 nM. It also displays high affinity for σ1 and σ2 receptors with Kis of 26 nM and 34 nM, respectively. The pharmacological actions of (±)-Vesamicol are attributed to the inhibition of acetylcholine transport into synaptic vesicles and subsequent quantal release of acetylcholine.
  • Potent vesicular acetylcholine transport inhibitor
  • Displays high affinity for σ1 and σ2 receptors
  • Inhibits acetylcholine transport into synaptic vesicles
  • Reduces subsequent quantal release of acetylcholine
  • Increases cytosolic acetylcholine levels in rat brain
  • Decreases vesicular acetylcholine levels in rat brain (except striatum)