Learn More
Medchemexpress LLC (±)-Vesamicol hydrochloride | 120447-62-3 | 99.8% | 50 MG
SDP
Catalog No. 5000413054
Encompass_Preferred
(±)-Vesamicol hydrochloride ((±)-AH5183 hydrochloride) is a potent vesicular acetylcholine transport inhibitor with a Ki of 2 nM. It also displays high affinity for σ1 and σ2 receptors with Kis of 26 nM and 34 nM, respectively.
  • Potent vesicular acetylcholine transport inhibitor (Ki of 2 nM)
  • High affinity for σ1 receptors (Ki of 26 nM)
  • High affinity for σ2 receptors (Ki of 34 nM)
  • For research use only
Catalog No. 50-004-13054 Supplier Medchemexpress LLC Supplier No. HYB1813A50MG
(±)-Vesamicol hydrochloride ((±)-AH5183 hydrochloride) is a potent vesicular acetylcholine transport inhibitor with a Ki of 2 nM. It also displays high affinity for σ1 and σ2 receptors with Kis of 26 nM and 34 nM, respectively.
  • Potent vesicular acetylcholine transport inhibitor (Ki of 2 nM)
  • High affinity for σ1 receptors (Ki of 26 nM)
  • High affinity for σ2 receptors (Ki of 34 nM)
  • For research use only