Learn More
Medchemexpress LLC HY-15601 5mg Medchemexpress, Vesatolimod CAS:1228585-88-3 Purity:>98%
SDP
Catalog No. 502030651
Encompass_Preferred
Medchemexpress, HY-15601 5mg Vesatolimod CAS:1228585-88-3 Vesatolimod (GS-9620) is a potent, selective and orally active agonist of Toll-Like Receptor (TLR7) with an EC50 of 291 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0651 Supplier Medchemexpress LLC Supplier No. HY156015MG
Medchemexpress, HY-15601 5mg Vesatolimod CAS:1228585-88-3 Vesatolimod (GS-9620) is a potent, selective and orally active agonist of Toll-Like Receptor (TLR7) with an EC50 of 291 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.