Learn More
TARGETMOL CHEMICALS INC VESNARINONE 10MG
SDP
Catalog No. 502576939
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Vesnarinone (Arkin) (INN) is a mixed phosphodiesterase 3 inhibitor and ion-channel modifier that has modest dose-dependent positive inotropic activity but minimal negative chronotropic activity. It also suppresses cell proliferation and induces apoptosis by inducing the expression of p21 an inhibitor of cyclin-dependent kinase activity in p53-mediated cell cycle arrest. purity: 98%
Catalog No. 50-257-6939 Supplier TARGETMOL CHEMICALS INC Supplier No. T346510MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Vesnarinone (Arkin) (INN) is a mixed phosphodiesterase 3 inhibitor and ion-channel modifier that has modest dose-dependent positive inotropic activity but minimal negative chronotropic activity. It also suppresses cell proliferation and induces apoptosis by inducing the expression of p21 an inhibitor of cyclin-dependent kinase activity in p53-mediated cell cycle arrest. purity: 98%