Learn More
TARGETMOL CHEMICALS INC VESNARINONE HCL 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T3465L10MG
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Vesnarinone HCl (OPC-8212 HCl) is an orally active phosphodiesterase 3 (PDE3) inhibitor.Vesnarinone HCl modulates calcium and potassium ions increasing calcium flux and decreasing potassium flux.Vesnarinone HCl is a new positive inotropic compound that enhances myocardial contractility and can be be used in heart failure studies.
Catalog No. 50-257-6937