Learn More
Perkin Elmer US LLC Vespel Ferrule
Catalog No. 502111787
Encompass_Preferred
Vespel ferrules are not reusable and not recommended for use with fused silica capillary columns. Quantity: Package of 10 Nut Size: 1/16 inch Ferrule Inner Diameter: 1/16 inch
Catalog No. 50-211-1787 Supplier Perkin Elmer US LLC Supplier No. L1003028
Vespel ferrules are not reusable and not recommended for use with fused silica capillary columns. Quantity: Package of 10 Nut Size: 1/16 inch Ferrule Inner Diameter: 1/16 inch