Learn More
Quality Lab Accessories 300mL Clear Class A 250mL Mark Outer Glass, Agilent/VanKel APP 3 compatible
Catalog No. NC1681797
Encompass
300mL Clear Class A 250mL Mark Outer Glass with Flat Bottom for Agilent/VanKel APP 3 Biodissolution.
Catalog No. NC1681797 Supplier Quality Lab Accessories Supplier No. GLA30003
300mL Clear Class A 250mL Mark Outer Glass with Flat Bottom for Agilent/VanKel APP 3 Biodissolution.