Learn More
Bellco Glass, Inc. Vessel 6 Side Arms/4 Baffles, 15L
SDP
Catalog No. 501944775
Encompass_Preferred
Suitable for all cell culture and fermentation. The vessel is compatible with bellco over head drives and magnetic stirrers. Supplied with 100mm center cap and 45mm caps and pouring ring. MADE IN THE USA 6 sidearms 4 baffles Includes: 100mm center cap 45mm screw Specifications:Material: Type 1 Class A Borosilicate Glass100mm Center NeckBaffles: 4Sidearms: 6OD x OAH: 295 x 460mm
Catalog No. 50-194-4775 Supplier Bellco Glass, Inc. Supplier No. 196415460
Suitable for all cell culture and fermentation. The vessel is compatible with bellco over head drives and magnetic stirrers. Supplied with 100mm center cap and 45mm caps and pouring ring. MADE IN THE USA 6 sidearms 4 baffles Includes: 100mm center cap 45mm screw Specifications:Material: Type 1 Class A Borosilicate Glass100mm Center NeckBaffles: 4Sidearms: 6OD x OAH: 295 x 460mm