Learn More
Chemglass Life Sciences Vessel, Bio Reactor, 3L, Low Profile, Graduated, 100-440 GPI Thread CN, (4) Vertical GL-45 SN's
SDP
Catalog No. 501532130
Encompass_Preferred
Vessel has four vertical GL-45 threaded side necks and a 100mm threaded center neck.The 3 and 6 liter vessels are low profile to maximize incubator space.Supplied complete with (4) solid GL-45 caps and (4) pouring rings.
Catalog No. 50-153-2130 Supplier Chemglass Life Sciences Supplier No. CLS140403
Vessel has four vertical GL-45 threaded side necks and a 100mm threaded center neck.The 3 and 6 liter vessels are low profile to maximize incubator space.Supplied complete with (4) solid GL-45 caps and (4) pouring rings.