Learn More
Chemglass Life Sciences Vessel, Bio Reactor, 8L, Graduated, 100-440 GPI Thread CN, (4) Vertical GL-45 SN's
SDP
Catalog No. 501532131
Encompass_Preferred
Vessel has four vertical GL-45 threaded side necks and a 100mm threaded center neck.The 3 and 6 liter vessels are low profile to maximize incubator space.Supplied complete with (4) solid GL-45 caps and (4) pouring rings.
Catalog No. 50-153-2131 Supplier Chemglass Life Sciences Supplier No. CLS140408
Vessel has four vertical GL-45 threaded side necks and a 100mm threaded center neck.The 3 and 6 liter vessels are low profile to maximize incubator space.Supplied complete with (4) solid GL-45 caps and (4) pouring rings.