Learn More
Fine Science Tools VESSEL CANNULATION FORCEPS 1MM
SDP
Catalog No. NC1575246
Encompass
Product: Dumont Vessel Cannulation Forceps - 1mm OD/11cm/ Tip Shape: Angled/ Tubing Size: 1mm OD/ Alloy: Inox/ Length: 11cm/ Unit: Each
Catalog No. NC1575246 Supplier Fine Science Tools Supplier No. 1128211
Product: Dumont Vessel Cannulation Forceps - 1mm OD/11cm/ Tip Shape: Angled/ Tubing Size: 1mm OD/ Alloy: Inox/ Length: 11cm/ Unit: Each