Learn More
Stoelting™ Vessel Cannulation Forceps Dumont INOX
SDP
Catalog No. 10001145 Shop All Stoelting Co Products
Change view
Click to view available options
Size:
0.05 mm OD
0.35 mm OD
1 mm OD
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
10-001-145 0.35 mm OD
10-001-146 0.05 mm OD
10-001-147 1 mm OD
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 10-001-145 Supplier Stoelting™ Supplier No. 5210065
Only null left
Add to Cart
Add to Cart
Stoelting™ Vessel Cannulation Forceps are useful for introducing a small plastic tube into a blood vessel of almost equal size.
Hollowed jaws hold tubing securely without deforming it and the tip is guided into the vessel opening while preventing damage from unwanted movement.
Specifications
For Use With (Application) Neuroscience
Size 0.35 mm OD