Learn More
Fine Science Tools S&T Vessel Cannulation Forceps, Angled, 11 cm
SDP
Catalog No. NC9092911
Encompass
Product: S&T Vessel Cannulation Forceps - 0.5-1mm OD/11cm/ Tip Shape: Angled/ Tubing Size: 0.5-1mm OD/ Alloy: Stainless Steel/ Length: 11cm/ Unit: Each
Catalog No. NC9092911 Supplier Fine Science Tools Supplier No. 0057411
Product: S&T Vessel Cannulation Forceps - 0.5-1mm OD/11cm/ Tip Shape: Angled/ Tubing Size: 0.5-1mm OD/ Alloy: Stainless Steel/ Length: 11cm/ Unit: Each