Learn More
Roboz Surgical VESSEL CANULATION FORCEPS
SDP
Catalog No. NC0241782
Encompass
These forceps are used to introduce fine plastic tubing into small blood vessels. The tips of these forceps are designed to securely grip the tubing without deforming or damaging it. They allow you to precisely guide the tubing into the blood vessel
Catalog No. NC0241782 Supplier Roboz Surgical Supplier No. RS4990
These forceps are used to introduce fine plastic tubing into small blood vessels. The tips of these forceps are designed to securely grip the tubing without deforming or damaging it. They allow you to precisely guide the tubing into the blood vessel