Learn More
World Precision Instrument Schwartz Vessel Clip, Strong Angle
SDP
Catalog No. 50822833
Encompass
WPI's Schwartz Vessel Clip is a surgical instrument for use in microvascular procedures, where small blood vessels and delicate tissues are involved. WPI Vessel Clips are designed to occlude fine blood vessels with precise control. These clips are made of German stainless steel and have 1.7 x 8mm smooth jaws with no teeth. These ligating vessel clips can tie off blood vessels or other tissues. The Schwartz vessel clips have a 45º angle, giving you greater visibility when placing the clip and access to deeper locations. Vessel clips are durable and withstand the stresses of surgery, ensuring that they remain securely in place throughout the procedure. Pinch the handle to release pressure and position the vessel clip. Then, release the handles to clamp the tissue or vessel securely. The size of the vessel clip you need can vary depending on the size of the vessel or tissue being occluded.
Catalog No. 50-822-833 Supplier World Precision Instrument Supplier No. 501788G
WPI's Schwartz Vessel Clip is a surgical instrument for use in microvascular procedures, where small blood vessels and delicate tissues are involved. WPI Vessel Clips are designed to occlude fine blood vessels with precise control. These clips are made of German stainless steel and have 1.7 x 8mm smooth jaws with no teeth. These ligating vessel clips can tie off blood vessels or other tissues. The Schwartz vessel clips have a 45º angle, giving you greater visibility when placing the clip and access to deeper locations. Vessel clips are durable and withstand the stresses of surgery, ensuring that they remain securely in place throughout the procedure. Pinch the handle to release pressure and position the vessel clip. Then, release the handles to clamp the tissue or vessel securely. The size of the vessel clip you need can vary depending on the size of the vessel or tissue being occluded.