Learn More
World Precision Instrument Vessel Clip, Full Curve
SDP
Catalog No. 50822829
Encompass
WPI's Full Curve Vessel Clip is a surgical instrument for use in microvascular procedures, where extremely small blood vessels and delicate tissues are involved. WPI Vessel Clips are designed to occlude fine blood vessels with precise control. These clips are made of German stainless steel and have 1 x 6mm jaws. These ligating vessel clips can tie off blood vessels or other tissues. Vessel clips are durable and withstand the stresses of surgery, ensuring that they remain securely in place throughout the procedure. Pinch the handle to release pressure and position the vessel clip. Then, release the handles to clamp the tissue or vessel securely. The size of the vessel clip you need can vary depending on the size of the vessel or tissue being occluded. These vessel clips are manufactured from German grade stainless steel, and they are suitable for autoclaving or chemical sterilization. Always clean surgical instruments thoroughly and promptly.
Catalog No. 50-822-829 Supplier World Precision Instrument Supplier No. 501784G
WPI's Full Curve Vessel Clip is a surgical instrument for use in microvascular procedures, where extremely small blood vessels and delicate tissues are involved. WPI Vessel Clips are designed to occlude fine blood vessels with precise control. These clips are made of German stainless steel and have 1 x 6mm jaws. These ligating vessel clips can tie off blood vessels or other tissues. Vessel clips are durable and withstand the stresses of surgery, ensuring that they remain securely in place throughout the procedure. Pinch the handle to release pressure and position the vessel clip. Then, release the handles to clamp the tissue or vessel securely. The size of the vessel clip you need can vary depending on the size of the vessel or tissue being occluded. These vessel clips are manufactured from German grade stainless steel, and they are suitable for autoclaving or chemical sterilization. Always clean surgical instruments thoroughly and promptly.