Learn More
World Precision Instrument Vessel Clips, 10g Pressure, Pkg of 5
SDP
Catalog No. 50822328
Encompass
Vessel clips are a surgical instrument used to clamp blood vessels during surgical procedures to temporarily occlude (block) blood flow in a specific area. These clips are made of metal and come in a variety of sizes. These ligating vessel clips can tie off blood vessels or other tissues. The size of the vessel clip you need can vary depending on the size of the vessel or tissue being occluded. These are available with 0.8x5mm (10g pressure), 1x6mm (15g pressure), 1.5x5mm (30g pressure) and 1.5x8mm (60g pressure) jaws. They come in packages of 5 clips. These vessel clips are manufactured from surgical grade stainless steel, and they are suitable for autoclaving or chemical sterilization. Always clean surgical instruments thoroughly and promptly.
Catalog No. 50-822-328 Supplier World Precision Instrument Supplier No. 15911
Vessel clips are a surgical instrument used to clamp blood vessels during surgical procedures to temporarily occlude (block) blood flow in a specific area. These clips are made of metal and come in a variety of sizes. These ligating vessel clips can tie off blood vessels or other tissues. The size of the vessel clip you need can vary depending on the size of the vessel or tissue being occluded. These are available with 0.8x5mm (10g pressure), 1x6mm (15g pressure), 1.5x5mm (30g pressure) and 1.5x8mm (60g pressure) jaws. They come in packages of 5 clips. These vessel clips are manufactured from surgical grade stainless steel, and they are suitable for autoclaving or chemical sterilization. Always clean surgical instruments thoroughly and promptly.