Learn More
METTLER TOLEDO Vessel for EVA V1/V3 KF Titrator
Catalog No. 014388545 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Size:
Small
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
01-438-8545 Large
01-438-8546 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-438-8545 Supplier METTLER TOLEDO Supplier No. 30869300
Add to Cart
Add to Cart
Elevate your analysis with our specialized accessories for the EVA Karl Fischer V1 and V3 Titrators.
  • Accessories for the EVA Karl Fischer EVA V1 and V3 Titrators will simplify the handling of challenging samples, ensuring precise results.
  • Customize your Karl Fischer process for greater convenience and flexibility.
TRUSTED_SUSTAINABILITY
Specifications
Type Titration Vessel
Size Large