Learn More
Chemglass Life Sciences COMPLETE Flat Bottom Vessel with 2-Port Drip Tube Assembly
SDP
Catalog No. 50143731
Encompass
Our addition and sampling vessel with hose barbed 2-port drip tube assemblies are designed for addition or sampling of media, acid or base.Assembly is ideal for use with bio reactors where addition or sampling is required.The drip tube assemblies are constructed of 316L electropolished stainless steel and include a blue polypropylene GL32 open top cap with a white silicone gasket.Drip tubes have a finish of 20Ra or better.The vessel measures 32mm Diameter x 94mm OAL.
Catalog No. 50-143-731 Supplier Chemglass Life Sciences Supplier No. CLS1437B01
Our addition and sampling vessel with hose barbed 2-port drip tube assemblies are designed for addition or sampling of media, acid or base.Assembly is ideal for use with bio reactors where addition or sampling is required.The drip tube assemblies are constructed of 316L electropolished stainless steel and include a blue polypropylene GL32 open top cap with a white silicone gasket.Drip tubes have a finish of 20Ra or better.The vessel measures 32mm Diameter x 94mm OAL.