Learn More
OHAUS™ Vessel Holder for Microplate Vortex Mixer
Catalog No. 021061031 Shop All OHAUS Products
Change view
Click to view available options
Holds:
125 and 250 mL Erlenmeyer Flasks
500 mL / 1 L Flasks
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
02-106-1031 125 and 250 mL Erlenmeyer Flasks
02-106-1030 500 mL / 1 L Flasks
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 02-106-1031 Supplier OHAUS™ Supplier No. 30400218
Add to Cart
Add to Cart
This item is not returnable. View return policy
OHAUS™ Vessel Holder for Microplate Vortex Mixer secures Erlenmeyer Flasks; requires a Universal Holder
  • For use with Ohaus Microplate Vortex Mixer
  • Available in two different sizes
  • Also includes a non-slip mat
TRUSTED_SUSTAINABILITY
Specifications
Holds 125 and 250 mL Erlenmeyer Flasks
Product Type Flask Holder
Material Plastic, Rubber
For Use With (Equipment) Heavy Duty Vortex Mixer
Includes Non-Slip Grip Mat
Packaging Quantity 1 Ea.