Learn More
Mettler Toledo Vessel KC Brown
Catalog No. 01672591 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Type:
Titration Vessel
Catalog No. Type
01-672-591 Titration Vessel
1 options
Catalog No. 01-672-591 Supplier Mettler Toledo Supplier No. 30988519
Add to Cart
Add to Cart
Brown glass titration vessel to improve sample dissolution and extraction
  • Valuable for controlling reactions when using alcohol-free reagents.
  • Compatible with the EVA coulometric Karl Fischer Titrators C1X, C1D, C3X, and C3D.
TRUSTED_SUSTAINABILITY
Specifications
Type Titration Vessel