Learn More
Mettler Toledo Vessel KFC Clear
Catalog No. 01672590 Shop All METTLER TOLEDO Products
Catalog No. 01-672-590 Supplier Mettler Toledo Supplier No. 30988518
Add to Cart
Add to Cart
A clear vessel allows lab operators to visually monitor the sample and reagent mixture, compatible with the EVA series.
  • Vessel KFC Clear is a titration vessel made by Mettler Toledo, designed for coulometric Karl‐Fischer titration (KF).
  • Clear titration vessel to improve sample dissolution and extraction.
  • Compatible with the EVA Coulometric Karl Fischer Titrators C1X, C1D, C3X, and C3D.