Learn More
Mettler Toledo Vessel KFC TS
Catalog No. 01672589 Shop All METTLER TOLEDO Products
Catalog No. 01-672-589 Supplier Mettler Toledo Supplier No. 30988517
Add to Cart
Add to Cart
Thermostatable titration vessel designed for coulometric Karl Fischer titration (KF), compatible with the KF titrator lines
  • Thermostatable titration vessel designed to improve sample dissolution and extraction.
  • Part of the broader KF accessory ecosystem, allowing use as the titration cell when paired with a KFC titrator and matching generator electrode, sensor, or injection setup.
  • Compatible with the EVA coulometric Karl Fischer Titrators C1X, C1D, C3X, and C3D.