Learn More
Perkin Elmer US LLC Vessel Rack Holder
Catalog No. 502112679
Encompass_Preferred
Acrylic box for holding eight Titan 75mL, 40 bar standard pressure vessels. The 40 bar standard pressure vessels fit in the 16-position carousel. Note: The 100 mL High Pressure Vessels for the 8-position carousel will not fit in this box.
Catalog No. 50-211-2679 Supplier Perkin Elmer US LLC Supplier No. N3134094
Acrylic box for holding eight Titan 75mL, 40 bar standard pressure vessels. The 40 bar standard pressure vessels fit in the 16-position carousel. Note: The 100 mL High Pressure Vessels for the 8-position carousel will not fit in this box.