Learn More
Chemglass Life Sciences 1000mL Reaction Vessel, Cylindrical, Jacketed, 100mm Schott O-Ring Flange, Approx body OD x Height 97mm I.D. x 205mm Height, Hose Conn. Thread Size GL-14
SDP
Catalog No. 50873499
Encompass_Preferred
1000mL Reaction Vessel, Cylindrical, Jacketed, 100mm Schott O-Ring Flange, 97mm I.D. x 205mm Height, Hose Conn. Thread Size GL-14. Cylindrical, jacketed, reaction vessel constructed with a flange having a groove formed in the sealing surface of the flange. Flange is also ground to permit a greased seal with any of the lids CG-1940 through CG-1946. Cooling jacket is equipped with two removable hose connections to reduce the risk of injury while attaching or detaching flexible tubing. Supplied complete with two polypropylene hose connections and two screw caps of the listed size. For clamp see CG-141 or CG-141-T. For o-ring see CG-147.PLEASE NOTE: HIGH TEMPERATURE HOSE CONNECTIONS AND SCREW CAPS ARE AVAILABLE. SEE CG-195-H AND CG-198-H.
Catalog No. 50-873-499 Supplier Chemglass Life Sciences Supplier No. CG192602
1000mL Reaction Vessel, Cylindrical, Jacketed, 100mm Schott O-Ring Flange, 97mm I.D. x 205mm Height, Hose Conn. Thread Size GL-14. Cylindrical, jacketed, reaction vessel constructed with a flange having a groove formed in the sealing surface of the flange. Flange is also ground to permit a greased seal with any of the lids CG-1940 through CG-1946. Cooling jacket is equipped with two removable hose connections to reduce the risk of injury while attaching or detaching flexible tubing. Supplied complete with two polypropylene hose connections and two screw caps of the listed size. For clamp see CG-141 or CG-141-T. For o-ring see CG-147.PLEASE NOTE: HIGH TEMPERATURE HOSE CONNECTIONS AND SCREW CAPS ARE AVAILABLE. SEE CG-195-H AND CG-198-H.