Learn More
Chemglass Life Sciences 3L Reaction Vessel, Jacketed, 150mm Schott O-Ring Flange, GL-14 Inlet/Outlet, CG-511-02 Drain Valve
SDP
Catalog No. 501531278
Encompass_Preferred
3L Reaction Vessel, Jacketed, 150mm Schott O-Ring Flange, GL-14 Inlet/Outlet, CG-511-02 Drain Valve. Similar to CG-1930, but with morton indentations to increase agitation during stirring. Supplied complete with two polypropylene hose connections and two screw caps. For clamp see CG-141 or CG-141-T. For o-ring see CG-147. Please Note: Morton Style Reactors/Flasks Not Suitable For Vacuum
Catalog No. 50-153-1278 Supplier Chemglass Life Sciences Supplier No. CG193126
3L Reaction Vessel, Jacketed, 150mm Schott O-Ring Flange, GL-14 Inlet/Outlet, CG-511-02 Drain Valve. Similar to CG-1930, but with morton indentations to increase agitation during stirring. Supplied complete with two polypropylene hose connections and two screw caps. For clamp see CG-141 or CG-141-T. For o-ring see CG-147. Please Note: Morton Style Reactors/Flasks Not Suitable For Vacuum