Learn More
Chemglass Life Sciences 3L Reaction Vessel, Jacketed, 150mm Schott O-Ring Flange, GL-14 Inlet/Outlet, CG-511-02 Drain Valve
SDP
Supplier:  Chemglass Life Sciences CG193026
Encompass_Preferred
3L Reaction Vessel, Jacketed, 150mm Schott O-Ring Flange, GL-14 Inlet/Outlet, CG51102 Drain Valve. Similar to CG1924, but with our 0-20mm CG511 sidearm valve. All valves are designed to seal flush with the bottom of the flask avoiding the dead space normally associated with stopcock drains. This feature permits total mixing & draining of the contents of the reactor. The CG510 is recommended for solution based reactions while the CG911A and CG511 are recommended for reactions containing solids. Reaction vessel is constructed with a flange having a groove formed in the sealing surface of the flange. Flange is also ground to permit a greased seal with any lids CG1940 through CG1946. Supplied complete with two polypropylene hose connections and two screw caps(Polypropylene hose connections with a Viton rubber seal for working temperatures up to max +110°C.) For clamp see CG141 or CG141T. For o-ring see CG147.
Catalog No. 50-873-502