Learn More
Chemglass Life Sciences 20000ml Reaction Vessel, Spherical, Morton, 150mm Schott O-Ring Flange, 350mm O.D. x 420mm Height
SDP
Catalog No. 50873507
Encompass_Preferred
22000ml Reaction Vessel, Spherical, Morton, 150mm Schott O-Ring Flange, 350mm O.D. x 420mm Height. Spherical shaped reaction vessel constructed with a flange having a groove formed in the sealing surface of the flange. Indentations increase agitation of the contents of the vessel during stirring. Flange is also ground to permit a greased seal with any of the lids CG-1940 through CG-1946. Vessel will fit standard heating mantles CG-10000 through CG-12502. For clamp see CG-141 or CG-141-T. For o-ring see CG-147.Please Note: Morton Style Reactors/Flasks Not Suitable For Vacuum
Catalog No. 50-873-507 Supplier Chemglass Life Sciences Supplier No. CG193405
22000ml Reaction Vessel, Spherical, Morton, 150mm Schott O-Ring Flange, 350mm O.D. x 420mm Height. Spherical shaped reaction vessel constructed with a flange having a groove formed in the sealing surface of the flange. Indentations increase agitation of the contents of the vessel during stirring. Flange is also ground to permit a greased seal with any of the lids CG-1940 through CG-1946. Vessel will fit standard heating mantles CG-10000 through CG-12502. For clamp see CG-141 or CG-141-T. For o-ring see CG-147.Please Note: Morton Style Reactors/Flasks Not Suitable For Vacuum