Learn More
Perkin Elmer US LLC Seal for Pressure Vessel M & H
Catalog No. 509044494
Encompass_Preferred
Lip type seal made of PTFE-TFM with venting system. Fits both pressure vessels of the Rotor 16, vessel MF100 and HF100.
Catalog No. 50-904-4494 Supplier Perkin Elmer US LLC Supplier No. N3141015
Lip type seal made of PTFE-TFM with venting system. Fits both pressure vessels of the Rotor 16, vessel MF100 and HF100.