Learn More
Ace Glass, Inc. Vessel, tapered, 2000mL, #36 center, (3) 24/40 sides
SDP
Catalog No. 50977133
Encompass_Preferred
Vessel, tapered, 2000mL, #36 center, (3) 24/40 sides
Catalog No. 50-977-133 Supplier Ace Glass, Inc. Supplier No. 983321
Vessel, tapered, 2000mL, #36 center, (3) 24/40 sides