Learn More
Ace Glass, Inc. Vessel, ultrasonic, 6-10mL, #25 center, (2)14/20 sides, body only
SDP
Catalog No. 50977138
Encompass_Preferred
Vessel, ultrasonic, 6-10mL, #25 center, (2)14/20 sides, body only
Catalog No. 50-977-138 Supplier Ace Glass, Inc. Supplier No. 984304
Vessel, ultrasonic, 6-10mL, #25 center, (2)14/20 sides, body only