Learn More
Lakeland Industries Vest
Supplier:  Lakeland Industries CV56
Encompass_Preferred
Pairing a cooling vest with your protective workwear can help combat the effects of heat stress in high hazard environments. CV56 features adjustable waist and shoulder straps with space to insert cooling packs. Made with a Banox FR cotton material.
Catalog No. 50-284-0461