Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS Vesta-Bac™ HB Ready-To-Use One-Step Disinfectant

Catalog No. 19415163 Shop All STERIS Corporation Products
Change view
Click to view available options
Volume (English):
22 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
19-415-163 22 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-415-163 Supplier STERIS Supplier No. 162377
Add to Cart
Add to Cart

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Vesta-Bac™ HB Ready-To-Use One-Step Disinfectant is a ready-to-use, quaternary ammonium chloride (quat)-based disinfectant spray with a broad range of activity against viruses that is intended to be used in laboratories and veterinary clinics.

Features
  • Virucidal
  • Ready-to-use
  • Multi-purpose
  • Serum effective
  • Recommended for use on floors, walls, metal surfaces, stainless-steel surfaces, glazed ceramic tile and other hard non-porous environmental surfaces
Benefits
  • Virucidal activity against HBV (Hepatitis B), HIV-1 virus (associated with AIDS)
  • Saves time and labor
  • Cleans, deodorizes and disinfects
  • Can also be used to control mold and mildew
  • Effective against Gram-negative and Grampositive bacteria, and pathogenic fungi
  • Bactericidal, fungicidal and virucidal in the presence of organic soil (5% bovine serum)

Specifications

Form Liquid Spray
pH 11
Appearance Light Green
Volume (English) 22 oz.
Volume (Metric) 650 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.