Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS Vesta-Syde™ SQ 64 RTU Non-Sterile Disinfectant

Catalog No. 17463005 Shop All STERIS Corporation Products
Change view
Click to view available options
Volume (English):
1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
17-463-005 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-463-005 Supplier STERIS Supplier No. 634208
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A phosphate-free, one-step cleaner and disinfectant formulated for use on hard, non-porous surfaces

  • Ready to Use Quaternary Ammonium Disinfectant
  • Formulated for use on hard, non-porous surfaces
  • Made with same main ingredients as the Vesta-Syde SQ Disinfectant concentrate
  • Microbial control against bacteria, fungi, and viruses using AOAC and EN protocols
  • At a use-dilution rate of 1.6% v/v (1:64), no mixing or activation is required for convenient and consistent use
  • Solution can be applied manually or sprayed
  • Low residue formulation to maintain visibly clean surfaces with minimal rinsing
  • Active ingredients notified according to the Biocide Product Directive and registered with the US EPA
  • Manufactured in accordance with the cGMP with complete lot traceability

Specifications

Product Type Quaternary Ammonium Disinfectant
Composition Phosphate-free
Concentration 1.6% v/v (1:64)
For Use With (Application) Microbial Control Against Bacteria, Fungi, and Viruses
Quantity 4/Cs.
Volume (English) 1 gal.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.