Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS Vesta-Syde™ SQ Disinfectant

Catalog No. 19415139 Shop All STERIS Corporation Products
Change view
Click to view available options
Volume (English):
1 gal.
55 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
19-415-139 1 gal.
17-463-039 55 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-415-139 Supplier STERIS Supplier No. 633508
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Quaternary ammonium disinfectant is an excellent fungicidal, non-sterile concentrate

  • Made specifically for use on hard non-porous surfaces
  • At a use-dilution rate of 0.8% v/v (1:128), the solution can be applied manually or sprayed
  • Fungicidal efficacy can support reduced sporicide use to control mold contamination
  • Formulated with biodegradable surfactants that provide excellent cleaning ability
  • Microbial control against bacteria, fungi, and viruses proven using AOAC and EN protocols
  • Active ingredients notified according to the Biocide Product Directive and registered with the US EPA
  • Manufactured in accordance with the cGMP with complete lot traceability

Specifications

Product Type Quaternary Ammonium Disinfectant
Form Concentrate
Sterility Non-sterile
Appearance Yellow
Certifications/Compliance Certificate of analysis
Concentration 0.8% v/v (1:128)
For Use With (Application) Microbial control against bacteria, fungi and viruses in cleanrooms
For Use With (Equipment) Mops and wipers
Quantity 4/Cs.
Volume (English) 1 gal.
Volume (Metric) 3.79 L
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.