Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS™ Vesta-Syde SQ Quaternary Ammonium Disinfectant

Catalog No. 19999573 Shop All STERIS Corporation Products
Change view
Click to view available options
Volume (English):
4 x 1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
19-999-573 4 x 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-999-573 Supplier STERIS™ Supplier No. 634008
Add to Cart
Add to Cart

STERIS' Vesta-Syde SQ Quaternary Ammonium Disinfectant is the non-sterile version of the Vesta-Syde SQ family of disinfectants made specifically for use on hard non-porous surfaces.Vesta-Syde SQ Disinfectant contains a formulation of quaternary ammonium compounds, surfactants and chelants.

Vesta-Syde SQ Disinfectant is specifically formulated for the routine decontamination of hard, non-porous surfaces. Vesta-Syde SQ Disinfectant contains a formulation of quaternary ammonium compounds, surfactants and chelants, and is available in multiple packaging configurations. At a use dilution rate of 0.78% v/v (1:128), Vesta-Syde SQ Disinfectant kills a broad spectrum of common bacteria and fungi including Aspergillus niger.

Features

  • Excellent fungicidal efficacy.
  • Broad spectrum efficacy against bacteria, fungi, and viruses using AOAC and EN protocols.
  • Superior material compatibility.
  • Formulated with biodegradable surfactants.
  • Active ingredients notified according to the Biocide Product Directive and registered with the US EPA.
  • Sterile version is gamma irradiated and each lot USP sterility tested.
  • Manufactured in accordance with cGMP with complete lot traceability.

Benefits

  • Can support reduced sporicide use to control mold contamination.
  • Confirms efficacy against many common microbes.
  • Allows for high frequency use on most surfaces.
  • Reduces environmental impact of disposal while maintaining excellent cleaning ability.
  • Permits global harmonization of disinfectants.
  • Facilitates introduction into critical areas.
  • Supports quality assurance and quality control.
  • Available in multiple packaging configurations.

Specifications

Product Type Cleaner/Disinfectant
Form Clear Liquid
pH 11.7 (Undiluted)
Scent Unscented
Sterility Non-sterile
Composition Quaternary Ammonium
Volume (English) 4 x 1 gal.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.